Learn More
VEE GEE ASTM Specific Gravity Hydrometer
Product Code. p-7250817
Click to view available options
ASTM Number:
111H
112H
113H
114H
115H
116H
117H
118H
119H
120H
125H
126H
127H
128H
129H
130H
131H
132H
133H
134H
Unit Size:
Each
Product Code. 17896583
Brand: VEE GEE 67117H
Only null left
Add to basket
Add to basket
The ASTM Specific Gravity hydrometer is manufactured to the specifications of the American Society for Testing and Materials.
  • Approx. Length: 330 mm
  • Calibrated for use at 15.5°C/60°F
Specifications
Type Specific Gravity Hydrometer
ASTM Number 117H
Increments 0.0005
Length (Metric) 33 cm
Description ASTM Specific Gravity Hydrometer, ASTM 117H, 1.300 to 1.350
Material Glass
Specific Gravity Range 1.450 to 1.500
Temperature of Standardization 15.5°C (60°F)