missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hecht Karl™ Assistent™ Amber Reagent Dropping Bottles without Closure
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
Specifications
Specifications
Capacity (Metric) | 10 mL |
Color | Amber |
For Use With (Application) | Hydrochloric acid |
Material | Glass |
Product Type | Dropping Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction