Learn More
Tocris Bioscience™ Asenapine maleate
Non-selective 5-HT, dopamine, adrenalin and histamine receptor antagonist; antipsychotic agent
€ 164.00 - € 642.00
Specifications
Purity | >98% |
---|---|
Product Type | Asenapine maleate |
Molecular Weight (g/mol) | 401.84 |
Inhibitors | Asenapine maleate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702544
|
Tocris Bioscience™
3737/10 |
10 mg |
€ 164.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712514
|
Tocris Bioscience™
3737/50 |
50 mg |
€ 642.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Novel psychopharmacologic agent. Displays antagonist activity at 5-HT, dopamine, noradrenalin and histamine receptor subtypes (pKi values are 8.60, 8.40, 10.15, 9.75, 10.46, 8.84, 9.60, 9.94, 8.85, 8.90, 8.84, 9.38, 8.95, 8.93, 8.9, 9.49, 8.91, 9.00 and 8.21 for 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT5A, 5-HT6, 5-HT7, D1, D2L, D2S, D3, D4, α1A, α2A, α2B, α2C, H1 and H2 receptors respectively). Displays no appreciable affinity for muscarinic receptors. Exhibits potent activity in animal models predictive of antipsychotic efficacy.Specifications
>98% | |
Asenapine maleate | |
401.84 | |
Asenapine maleate |
For Research Use Only