Learn More
Thermo Scientific™ ART™ Barrier Specialty Pipette tips
Perform pipetting accurately and consistently with these barrier extended length pipette tips, which seal when exposed to potential contaminants.
Brand: Thermo Scientific™ 2140
Description
Perform applications consistently and accurately by using Thermo Scientific™ ART™ Barrier Specialty Pipette Tips, which seal when exposed to potential contaminants, trapping them inside the barrier. ART barrier specialty tips are designed to aid in pipetting for a variety of applications. Extended length, Thermo Scientific™ REACH™ tips provide an extra level of contamination control.
Features:
- Sterile tips are certified free from RNase, DNase and pyrogen
- Available in sterile, nonsterile and in various packaging options
- Manufactured according to the industry's highest quality standard
Designed to Aid in Ppipetteting for a Variety of Applications:
Extended length: to prevent contamination by keeping the pipettete shaft from entering the sample vessel
Gel loading: for loading acrylamide or agarose gels
Wide orifice genomic: for genomic DNA, fragile cell lines and other viscous materials
Solvent safe: for acids, bases and aggressive organic solvents
Compatible with:
SoftFit™ pipette tips provide precise and consistent pipetteting with multichannel pipettes
Ergonomic pipette tips are designed for Gilson Pipetman™ pipettes and Thermo Scientific™ Finnpipette™ Focus pipettes

Specifications
ART 10 REACH, Filtered, Sterile, Lift-off Lid Rack | |
Filtered | |
ART 10 REACH | |
4.33cm | |
Lift-off Lid Rack | |
Universal Fit | |
Sterile and PCR Ready |
10 μL | |
Sterile | |
1.71 in. | |
DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free | |
Extended Length | |
ART |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.