Learn More
Antimony Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M, Fisher Chemical™
Product Code. 10641533
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Antimony Standard Metal Solution 1000ppm, for AAS, Solution HCl 1M
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number 134030
Specifications
Packaging HDPE plastic bottle
Quantity 100 mL
Color Colorless
Physical Form Liquid
compliance-icons
Product Identifier
  • Antimony solution 1000 ppm
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.