Learn More
TNFRSF17, Human, Matched Antibody Pair (AP41), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00000608-AP41.1Set
Product Code. 16171774
  • € 809.00 / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-TNFRSF17 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TNFRSF17, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TNFRSF17
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TNFRSF17.
BCM/BCMA/CD269
Mouse
RUO
608
Human
Liquid
ELISA
Biotin
TNFRSF17
TNFRSF17
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TNFRSF17 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TNFRSF17, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only