missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-RALGDS, HRAS Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0565.1Set
Includes: RALGDS rabbit purified polyclonal antibody (20μg); HRAS mouse purified polyclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the RALGDS protein, and the other against the HRAS protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
RALGDS | |
Monoclonal/Polyclonal | |
RALGDS | |
RALGDS | |
1 Set | |
Primary | |
Antibody pair set content: 1. RALGDS rabbit purified polyclonal antibody (20 ug) 2. HRAS mouse purified polyclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
FLJ20922/RGF/RalGEF | |
Mouse, Rabbit | |
RUO | |
5900/3265 | |
Human | |
RALGDS & HRAS Protein Protein Interaction Antibody Pair |