missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-NKD2 Matched Antibody Pair, Abnova™
Mached Pair Antibody
Brand: Abnova H00085409-AP21.1Set
Additional Details : Weight : 0.00010kg
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NKD2 (100μg); Detection antibody: mouse purified polyclonal anti-NKD2 (20μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
Host Species: Rabbit, Mouse
Cross Reactivity: Human
ELISA Pair
Specifications
NKD2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NKD2. | |
Naked2 | |
Mouse, Rabbit | |
1 Set | |
85409 | |
Human |
ELISA | |
Unconjugated | |
NKD2 | |
NKD2 | |
Affinity Purified | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-NKD2 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-NKD2 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |