Learn More
potassium voltage-gated channel, shaker-related subfamily, member 3, Mouse, Clone: 1D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KCNA3.
Brand: Abnova H00003738-M01.100ug
Description
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It belongs to the delayed rectifier class, members of which allow nerve cells to efficiently repolarize following an action potential. It plays an essential role in T-cell proliferation and activation. This gene appears to be intronless and it is clustered together with KCNA2 and KCNA10 genes on chromosome 1. [provided by RefSeq
Sequence: VPVIVSNFNYFYHRETEGEEQSQYMHVGSCQHLSSSAEELRKARSNSTLSKSEYMVIEEGGMNHSAFPQTPFKTGNSTATCTTNNNPNSCVNIKKIFTDVSpecifications
potassium voltage-gated channel, shaker-related subfamily, member 3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC035059 | |
KCNA3 | |
KCNA3 (AAH35059, 424 a.a. ∼ 523 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Western Blot | |
1D8 | |
Mouse monoclonal antibody raised against a partial recombinant KCNA3. | |
KCNA3 | |
HGK5/HLK3/HPCN3/HUKIII/KV1.3/MK3/PCN3 | |
Mouse | |
Affinity Purified | |
RUO | |
3738 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.