Learn More
EFNA3, Human, IP-WB Antibody Pair (PW1), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00001944-PW1.1Set
Additional Details : Weight : 0.02000kg
Product Code. 16034798
  • € 789.00 / 1 set
Estimated Shipment: 26-07-2024
to see stock.
Includes: Antibody pair for IP: mouse monoclonal anti-EFNA3 (300μg), Antibody pair for WB: rabbit polyclonal anti-EFNA3 (50μL)
Specifications
EFNA3
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
EFL2/EPLG3/Ehk1-L/LERK3
Mouse, Rabbit
RUO
1944
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
EFNA3
EFNA3
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-EFNA3 (300 ug) 2. Antibody pair for WB: rabbit polyclonal anti-EFNA3 (50 ul)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only