Learn More
CD40LG, Human, Matched Antibody Pair (AP15), Abnova™
To detect and quantify protein level of human
Brand: Abnova H00000959-AP15.1Set
Includes: Capture antibody: rabbit MaxPab affinity purified polyclonal anti-CD40LG (100μg) , Detection antibody: mouse monoclonal anti-CD40LG, IgG1 Kappa (20μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
CD40LG | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CD40LG. | |
CD154/CD40L/HIGM1/IGM/IMD3/T-BAM/TNFSF5/TRAP/gp39/hCD40L | |
Mouse, Rabbit | |
RUO | |
959 | |
Human | |
Liquid |
ELISA | |
Unconjugated | |
CD40LG | |
CD40LG | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CD40LG (100 μg) 2. Detection antibody: mouse monoclonal anti-CD40LG, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only