missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD147 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 345600
This item is not returnable.
View return policy
Description
CD147 Polyclonal antibody specifically detects CD147 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
CD147 | |
Polyclonal | |
Unconjugated | |
BSG | |
5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
682 | |
-20°C | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P35613 | |
BSG | |
Synthetic peptide derived from the N-terminal region of the human EMMPRIN protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |