missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, Unlabeled, Clone: M5E2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557152
This item is not returnable.
View return policy
Description
Specifications
| CD14 | |
| Monoclonal | |
| 0.5mg/mL | |
| LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| M5E2 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CD14 Protein | |
| 0.1 mg | |
| Immunology | |
| Canine, Cynomolgus, Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction