missing translation for 'onlineSavingsMsg'
Learn More

anti-CCNB1, FZR1 Protein Protein Interaction Antibody Pair, Abnova™

Brand:  Abnova DI0213.1Set

Product Code. 16161194


Includes: CCNB1 rabbit purified polyclonal antibody (20μg); FZR1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
This item is not returnable. View return policy

Description

Description

This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the CCNB1 protein, and the other against the FZR1 protein for use in in situ Proximity Ligation Assay.

Host Species: Rabbit, Mouse
Species Reactivity: Hu

In situ PLA

Specifications

Specifications

CCNB1
Monoclonal/Polyclonal
CCNB1
CCNB1
1 Set
Primary
Antibody pair set content: 1. CCNB1 rabbit purified polyclonal antibody (20 ug) 2. FZR1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Purified
In situ PLA
Unconjugated
CCNB
Mouse, Rabbit
RUO
891/51343
Human
CCNB1 & FZR1 Protein Protein Interaction Antibody Pair
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers