missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA514208
This item is not returnable.
View return policy
Description
C1QA Polyclonal Antibody for Western Blot, IHC (P), Flow
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
Specifications
| C1QA | |
| Polyclonal | |
| Unconjugated | |
| C1qa | |
| AI255395; C1q; C1QA; complement C1q A chain; complement C1q chain A; complement C1q subcomponent subunit A; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component C1q, A chain | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P02745 | |
| C1qa | |
| KLH conjugated synthetic peptide between 198-227 amino acids from the C-terminal region of human C1QA. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction