missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein H Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537840
This item is not returnable.
View return policy
Description
This antibody is predicted to react with dog based on sequence homology.
Apolipoprotein H Polyclonal antibody specifically detects Apolipoprotein H in Human samples. It is validated for Western Blot.Specifications
Apolipoprotein H | |
Polyclonal | |
Unconjugated | |
APOH | |
2GPI; activated protein C-binding protein; Anticardiolipin cofactor; APC inhibitor; APOH; Apo-H; Apolipoprotein H; apolipoprotein H (beta-2-glycoprotein I); B2G1; B2GP1; B2GPI; beta(2)GPI; beta-2-glycoprotein 1; Beta-2-glycoprotein I; beta-2-GPI; beta2-GPI; BG | |
Goat | |
Antigen affinity chromatography | |
RUO | |
350 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P02749 | |
APOH | |
Peptide with sequence C-QGERVKIQEKFKN, from the internal region of APOH | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |