missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ankyrin G Monoclonal Antibody (4G3F8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ 338800
This item is not returnable.
View return policy
Description
Ankyrin G Monoclonal Antibody for ELISA, ICC/IF, IP, Western Blot
Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.
Specifications
| Ankyrin G | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| G5E8K5, O70511, Q12955 | |
| ANK3 | |
| A synthetic peptide derived from the spectrin-binding domain of the human ankyrin G protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 4G3F8 | |
| Unconjugated | |
| ANK3 | |
| 2900054D09Rik; AI314020; ANK3; ANK-3; AnkG; ankyrin 3; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin G; ankyrin-3; Ankyrin-G; brain-specific ankyrin-G; LOW QUALITY PROTEIN: ankyrin-3; MRT37 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100353296, 11735, 288, 361833, 479215 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction