missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratoires Anios Anioxy-WS Detergent Disinfectant Spray
Broad spectrum disinfectant detergent for nonimmersible and non-invasive medical devices. (stethoscopes, pressure sensors, blood sugar testers, etc.)
Brand: Laboratoires Anios 1756573
This item is not returnable.
View return policy
Description
Disinfectant detergent spray:
- Clear liquid solution.
- pH at +20°C : ≈ 7,5.
- Keep away from heat.
- Stable at storage from +5°C to +25°C.
Specifications
Liquid | |
EN 1040, EN 1276, EN 13727, EN 14563, EN 1275, EN 13624, EN1650, NF T 72-190, EN 13704, EN 14561, NF T 72-190, EN 13697 | |
1 L |
7.5 | |
5°C to 25°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ANIOXY-SPRAY WS
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : ETHYL ALCOHOL, HYDROGEN PEROXIDE, AMINES, C12-14 (EVEN NUMBERED)-ALKYLDIMETHYL,N-OXIDES