missing translation for 'onlineSavingsMsg'
Learn More

Laboratoires Anios Aniosgel 85 NPC; Eighty-Five NPC Disinfecting Gel

Thixotropic hydroalcoholic gel for hygienic treatment and surgical disinfection by hand-rubbing.

Brand:  Laboratoires Anios 1644274


Product Code. 11841703

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Disinfecting gel for hydroalcoholic hand-rubbing:

  • Ethyl alcohol: 755 ml/l
  • Flash point: +23°C
  • Density at +20°C: approx. 0.862
  • pH at +20°C: approx. 5.5 (near to skin pH)
  • Viscosity at +20°C: from 1200 to 2500 mPa.s
  • Storage: from +5°C and +25°C
  • Aerobiocontamination study available on request
Specifications

Specifications

Aniosgel 85 NPC Disinfecting Gel
Viscous Liquid
5.25
75 mL
Colorless
EN 1040, EN 12054, EN 1500, EN 12791 and T 72–301, NF EN 1275
20 Pk.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ANIOSGEL 85 NPC

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403-Store in a well-ventilated place.

Supplemental information
  • MIXTURE LIST-Contains : ETHANOL, PROPAN-2-OL

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.