missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ancitabine hydrochloride, MedChemExpress
Click to view available options
Quantity:
200 mg
Unit Size:
200mg
Description
Ancitabine (hydrochloride) is an important antileukemia drugs.
Specifications
Specifications
| Chemical Name or Material | Ancitabine hydrochloride |
| CAS | 10212-25-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C9H12ClN3O4 |
| Quantity | 200 mg |
| Synonym | Cyclocytidine hydrochloride Cyclo-CMP hydrochloride Cyclo-C |
| Solubility Information | H2O : ≥ 50 mg/mL (191.09 mM) ∣DMSO : 25 mg/mL (95.54 mM; Need ultrasonic) |
| SMILES | OC[C@@H]1[C@@H](O)[C@@](O2)([H])[C@@](N(C2=N3)C=CC3=N)([H])O1.[H]Cl JjbwQ0byOjHpJ4gXWRYObRsaR41DaP== |
| Molecular Weight (g/mol) | 261.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction