Learn More
Abeba™ Anatom 32156 Shoes
Resperation absorbing special lining with silver ions. Steel toe-cap.
Brand:  Abeba™ 32156/39
 View more versions of this product
Product Code. 15649397
  • € 121.00 / 1 pair
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Description
  • CE EN ISO 20345:2011 S2 SRC
  • Smooth leather
  • Black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3580)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Suitable for heel and foot lift
  • Scratch-resistant coating in heel and toe area
Specifications
Specifications
CE EN ISO 20345:2011 S2 SRC
Leather
Slip-resistant
32156
Black
39
Polyurethane
Steel