Learn More
Hamilton™ Analytical Guard Cartridge Starter Kit
Brand: Hamilton™ 79368
This item is not returnable.
View return policy
Specifications
10 μm | |
Includes 1 holder, 2 cartridges | |
PEEK | |
PRP-X200 Analytical Guard Cartridge Starter Kit |
Product Content Correction
Product Title
Hamilton™ Analytical Guard Cartridge Starter Kit > Particle Size: 10μm