Learn More
Kern Sohn Analytical Balance ABT
Click to view available options
Capacity:
101 g
120 g
220 g
320 g
42 g, 120 g
82 g, 220 g
Unit Size:
Each
Description
- Automatic internal adjustment in the case of a change in temperature = 0,5°C or time-controlled every 4 h, guarantees high degree of accuracy and makes the balance independent of its location of use
- Dosage aid: High stability mode and other filter settings can be selected
- Simple recipe weighing and documenting with a combined tare/print function. In addition, the ingredients for the recipe are numbered automatically and printed out with their corresponding number and nominal weight
- Identification number: 4 digits, printed on calibration protocol freely programmable
- Printout of a GLP-compliant calibration report conveniently at the touch of a button
- Automatic data output to the PC/printer each time the balance is steady
- Large glass draught shield with 3 sliding doors for easy access to the items being weighed.
- Protective working cover included with delivery
Specifications
Specifications
| Capacity | 120 g |
| Readability | 0.1 mg |
| Linearity | 0.2 mg |
| Repeatability | 0.1 mg |
| Electrical Requirements | Universal Mains Adapter |