missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphotericin B, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
Specifications
Specifications
Type | Amphotericin B |
Physical Form | Powder |
Color | Yellow |
Quantity | 5 g |
IUPAC Name | (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19Z,21Z,23Z,25Z,27Z,29Z,31Z,33R,35S,36R,37S)-33-{[(2R,3S,4S,5S,6R)-4-amino-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid |
Formula Weight | 924.1 |
Chemical Name or Material | Amphotericin B, from Streptomyces sp. |
Safety and Handling

Product Identifier
- Amphotericin B
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Amphotericin B
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Amphotericin B, MP Biomedicals >
Spot an opportunity for improvement?Share a Content Correction