missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfamate, ≥98.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 131°C to 135°C |
Color | White |
Quantity | 100 g |
Linear Formula | NH4SO3NH2 |
UN Number | UN3077 |
Formula Weight | 114.12g/mol |
Percent Purity | ≥98.0% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Residue after Ignition | ≤0.10% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'