missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium fluoride, Puriss. p.a., ACS Reagent, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
1 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | NH4F |
MDL Number | MFCD00011423 |
UN Number | 2505 |
Formula Weight | 37.04g/mol |
Percent Purity | ≥98% |
Grade | ACS/puriss. p.a. |
Packaging | Plastic Bottle |
Residue after Ignition | ≤0.005% (as SO4) |
Chemical Name or Material | Ammonium Fluoride |
Sicurezza e movimentazione
Identificativo prodotto
- Ammonium fluoride
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ammonium fluoride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'