Learn More
WTW™ Ammonia and Ammonium Test Kits
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
Description
- For waste water, drinking water
- Chromate (chromium VI and total chromium) cell test kit
- Photometric test method
- This test is highly accurate and easy to use
Specifications
Specifications
| Analytes | Indophenol blue (Berthelot reaction) |
| For Use With (Application) | For waste water, drinking water, sea water |
| For Use With (Equipment) | PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers |
| Includes | Certificates |
| No. of Tests | 25 |
| Sufficient For | 25 Tests |
| Test Method | Photometric |
| Type | CombiCheck Ammonia (NH3) cell test kit |