Learn More
Aminoacylase 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11316-1-AP-150UL
 View more versions of this product
Product Code. 16818313
  • € 410.00 / 150µL
Description
ACY1 encodes a cytosolic, homodimeric, zinc-binding enzyme that catalyzes the hydrolysis of acylated L-amino acids to L-amino acids and an acyl group, and has been postulated to function in the catabolism and salvage of acylated amino acids. This gene is located on chromosome 3p21.1, a region reduced to homozygosity in small-cell lung cancer (SCLC), and its expression has been reported to be reduced or undetectable in SCLC cell lines and tumors. The amino acid sequence of human aminoacylase-1 is highly homologous to the porcine counterpart, and this enzyme is the first member of a new family of zinc-binding enzymes. Mutations in this gene cause aminoacylase-1 deficiency, a metabolic disorder characterized by central nervous system defects and increased urinary excretion of N-acetylated amino acids. Alternative splicing of this gene results in multiple transcript variants. Read-through transcription also exists between this gene and the upstream ABHD14A (abhydrolase domain containing 14A) gene. A related pseudogene has been identified on chromosome 18.
Specifications
Specifications
Aminoacylase 1
Polyclonal
Unconjugated
ACY1
ACY 1, ACY1, ACY1D, ACYLASE, Aminoacylase 1
Rabbit
Antigen Affinity Chromatography
RUO
109652, 300981, 95
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q03154, Q6AYS7, Q99JW2
ACY1
Aminoacylase 1 Fusion Protein Ag1813
150 μL
Primary
Human, Mouse, Rat