Learn More
Cytiva Amersham™ Hybond™ P 0.45μm PVDF Membrane
Product Code. p-4709761 Shop All Cytiva Products
Click to view available options
Quantity:
1 Roll/Pk.
10 Sheets/Pk.
25 Sheets/Pk.
5/Pk.
Dimensions (LxW):
100 x 100mm
140 x 160mm
150mm x 4m
300 x 600mm
300mm x 4m
80 x 90mm
Unit Size:
1 roll
10 sheets
25 sheets
5 sheets
Product Code. 15289894
Brand: Cytiva 10600069
Use for chemiluminescent and fluorescent detection methods for proteins of > Mr 20 000. Cytiva Life Sciences™ Amersham™ Hybond™ P 0.45μm PVDF Membrane is a 0.45 μm pore size hydrophobic membrane, with high physical strength, highly suitable for stripping and reprobing.
  • Protein binding capacity over over 200 μg/cm2 for easy signal detection
  • High protein retention even after harsh washing steps
  • High protein capture during transfer minimizes sample loss
  • Available in popular precut sizes for your application
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 Sheets/Pk.
Pore Size 0.45 μm
Dimensions (LxW) 140 x 160mm