missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ 4 to 20%, Tris-HCl, 1.4 mm, Protein Gel, 15-well
Separation Type: Denaturing, Native; For Use With: Amersham ECL Gel Box
Brand: Cytiva 28-9901-59
This item is not returnable.
View return policy
Description
Homogeneous and gradient gels for horizontal protein electrophoresis.
- Amersham™ ECL™ Gels have a shelf life of up to 12 months with no compromise in performance over time
- Gels are available in 15 and 10 well formats as either homogenous (10 and 12%) or gradient (4-12%, 8-16% and 4-20%)
- Compatible with standard Laemmli buffers
- Works well with standard protein detection protocols
- Amersham ECL Gel is cast without SDS, allowing the user to define the separation conditions by the composition of the running buffer and sample loading buffer
Specifications
Clear | |
4 to 20% | |
Tris-HCl | |
Protein | |
15-well | |
Polyacrylamide Gel Electrophoresis | |
4° to 8°C | |
20 μL | |
7.5 cm | |
3.15 in. | |
8 cm | |
7.5 cm |
Amersham ECL Gel Box | |
1.4 mm | |
Gradient Page | |
Denaturing, Native | |
Acrylamide/bis-acrylamide, %C = 2.6% (w/w), %T = Total concentration (w/v) of Acrylamide + bis-acrylamide 4 to 20% | |
10/Pk. | |
0.05 in. | |
75 mm | |
3.15 in. | |
80 mm | |
2.95 in. |