Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Product Code. 11565055 Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy2 bis-NHS Ester
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy5.5 bis-NHS Ester
Cy7 Bis NHS Ester
Cy7 bis-NHS Ester
Unit Size:
10mg
50mg
5mg
Pack of 5
Product Code. 11565055
Brand: Cytiva PA17004
Suitable for general labeling of amine residues
Order Info
Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used for high-throughput screening (HTS) applications
Quantity 10 mg
Product Type Cy7 Bis NHS Ester
Kit Contents Foil packs, each containing 5, 10 or 50mg dried dye
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Color Purple
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 2
Hazard Statement
  • H371-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cy7 (Bis NHS ester)