Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Shop All Cytiva Products
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Product Type:
Cy2 bis-NHS Ester
Cy3 bis-NHS Ester
Cy5 Bis NHS Ester
Cy5 bis-NHS Ester
Cy5.5 Bis NHS Ester
Cy5.5 bis-NHS Ester
Cy7 Bis NHS Ester
Cy7 bis-NHS Ester
Unit Size:
10mg
50mg
5mg
Pack of 5
Description
Order Info
Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
Specifications
Specifications
| For Use With (Application) | Suitable for general labeling of Amine residues |
| Quantity | 5 mg |
| Product Type | Cy7 bis-NHS Ester |
| Kit Contents | Foil packs, each containing 5, 10 or 50mg dried dye |
| Content And Storage | 2°C to 8°C |
Safety and Handling
Signal Word
- Warning
- Specific target organ toxicity Category 2