Learn More
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs
Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Brand: Cytiva PA22000
This item is not returnable.
View return policy
Description
- Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
- For use in ELISA, immunoprecipitation, and blotting applications.
- Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Order Info
Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.
Specifications
For the labelling of biological compounds with Cy2 Reactive Dye | |
Cy2 bis-Reactive Dye Pack | |
Labeling 1mg Protein or 30nM Amino groups | |
2°C to 8°C, protected from light | |
Yellow |
Each | |
Five foil packs each containing dried dye to label 1mg of Protein | |
896.95 | |
Cy2 bis-Reactive Dye |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
Product Content Correction
Product Title
Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs > Cy2 bis-Reactive Dye Pack