Learn More
Invitrogen™ MEGAscript™ T7 Transcription Kit
Product Code. p-4918894
Click to view available options
Product Type:
Kit with manual
T7 In Vitro Transcription kit
No. of Reactions:
200 Reactions
25
25 Reactions
40
40 Reactions
Unit Size:
25 reactions
Each
Product Code. 10600616
Brand: Invitrogen™ AMB13345
Edge
Includes
T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II, Nuclease-free Water
Ultra-high yield technology for in vitro transcription.
The MEGAscript T7 Kit is an ultra-high yield in vitro transcription kit. The high yields are achieved by modifying typical transcription reaction conditions so that very high nucleotide concentrations can be effectively used. The T7 Enzyme Mix and the 10X Reaction Buffer are specifically calibrated for each lot and RNA polymerase.
Using the MEGAscript T7 Kit
The MEGAscript T7 Kit contains in vitro transcription reaction components and a control template. The kit will yield a total of 3–5 mg of RNA (approximately 100 μg of RNA or more per reaction) from the control template supplied with the kit. This corresponds to 400–650 moles of RNA for each mole of template. Smaller templates typically yield a lower mass and a higher molar yield of product. Novel transcription reaction conditions and a patented, high-yield technology allow the synthesis of 10–50 times the amount of RNA produced by conventional transcription reactions.
Applications
The MEGAscript T7 Kit is intended for the synthesis of large amounts of unlabeled or low specific activity RNA for a variety of uses, including in vitro translation, antisense/microinjection studies, and isolation of RNA binding proteins. In large-scale transcription reactions, the concentration of all four nucleotides is high, well above the Km for the enzyme. The MEGAscript T7 Kit typically yields over 10 times more RNA than conventional in vitro transcription reactions. The kit is not recommended for synthesis of high specific activity probes.
Accessory products
MessageAmp aRNA amplification kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use the MEGAclear™ Kit (Cat. No. AM1908) and MEGAclear-96 Kit (Cat. No. AM1909).
RNA yield: 100 μg
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 200 Reactions
Product Line Ambion, MEGAscript
Product Type T7 In Vitro Transcription kit
Quantity 200 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield 100 μg
Starting Material PCR Products, Linearized DNA plasmid