Learn More
Lenz Laborglasinstrumente™ Amber Borosilicate Glass Separating Funnel with PTFE Plug
Product Code. 11983441
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 2
Product Code. 11983441
Brand: Lenz Laborglasinstrumente™ 04008149
Only null left
Add to basket
Add to basket
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Amber Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into solvent phases of specific densities.
  • With NS socket and PE stopper
  • Without scale
  • With PTFE plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3;
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Includes PTFE plug and screw thread retaining device
Joint Number 29/32 NS Socket
Product Type Separatory Funnel