Learn More
Sartorius™ Aluminum Special Weight With Factory Certificate
Product Code. 10223281 Shop All Sartorius Products
Click to view available options
:
Mass: 2 x 50g
Product Code. 10223281
Brand: Sartorius™ YSS4500
This item requires a price update prior to being ordered.
Request a Price for this item here.
Accessory weight set for air density determination, for ME series balances
  • For ME series
  • For air density determination
TRUSTED_SUSTAINABILITY
Specifications
Description Weight set for aie density determination
For Use With (Equipment) ME series balances