Learn More
Korff™ Aluminum Small Rolls
Click to view available options
Width (Metric):
300mm
450mm
Description:
Foil Gauge: 0.015mm
Foil Gauge: 0.030mm
Unit Size:
1 roll
10m
Specifications
Specifications
| Description | Foil Gauge: 0.015mm |
| Product Type | Small Rolls |
| Width (Metric) | 300mm |
| Material | Aluminum |