Learn More
Brady™ Aluminum Sign: Evacuation assembly point - ISO 7010
Product Code. p-7159753
Click to view available options
Height (Metric):
400 mm
600 mm
Width (Metric):
400 mm
600 mm
Unit Size:
Each
Product Code. 15684197
Brand: Brady™ 138853
Only null left
Add to basket
Add to basket
Improve workplace safety with proper signage and markings. This Brady™ aluminum sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • High performance material for maximum durability
  • Clearly visible and easy to understand
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number E/E007/NT-ALU-400x400/1-B
Material Aluminum
Height (Metric) 400 mm
Thickness 1.016 mm
Width (Metric) 400 mm
Temperature Range (Metric) -40°C to 140°C
Length (Metric) 400 mm
Shape Square
Languages Global
Color White on Green