missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum with Polyester Overlaminate Stickers for the Transport of Dangerous Goods

Product Code. 11764914
Click to view available options
Material:
Aluminum
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11764914

Brand: Brady™ 258971

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Material Aluminum
Quantity 1 Ea.
Height (Metric) 297 mm
Thickness 1.016 mm
Width (Metric) 297.00 mm
Temperature Range (Metric) -40 °C-140 °C
Shape Rhomboid
Displayed Text 6
Color Black on White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.