Learn More
Brady™ Aluminum with Polyester Overlaminate Mandatory Signs
Product Code. 11774894
Click to view available options
Diameter (Metric):
100 mm
200 mm
315 mm
Displayed Text:
Commandment Icon- Overcoat required
Protective Gloves
Use Head Protection
Use breathing protection
Use face shield
Use full protective
Use overshoes
Use protective hood
Using Foot
Wear protective apron
missing translation for 'unitSize'
Each
Product Code. 11774894
missing translation for 'mfr': Brady™ 257924
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance ISO 7010
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
Diameter (Metric) 100 mm
For Use With (Application) Indoor and outdoor safety signs
Thickness 0.271 mm
Temperature Range (Metric) -40°C to 140°C
Shape Round
Displayed Text Use overshoes
Product Type Mandatory Sign
Color White on Blue