missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitrate Nonahydrate, ≥98%, ACS Reagent, Honeywell Fluka™

Product Code. 15692370
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73°C
Color White
pH 2.5 to 3.5 at 20°C
Quantity 100 g
Linear Formula Al(NO3)3 · 9H2O
UN Number NONH for all modes of transport
Formula Weight 375.13g/mol
Percent Purity ≥98%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Crystals
Chemical Name or Material Aluminum Nitrate Nonahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'