Learn More
Nickel Electro™ Aluminum Lids for Aluminum Basin
Shop All Nickel-Electro Products
Click to view available options
Diameter (Metric):
60 mm
70 mm
75 mm
90 mm
200 mm
Unit Size:
Pack of 5
Description
Nickel Electro™ Aluminun Lid for aluminum Basin is crevice free and one piece construction.
- Aluminum
- Seamless
- One piece construction
- Crevice free
- For use with aluminum basin
Specifications
Specifications
| Material | Aluminum |
| Type | Lid |
| Height (Metric) | 5 mm |
| Diameter (Metric) | 70 mm |