Learn More
Welch™ Aluminum KF Inner Centering with Buna N O-ring
Shop All Welch Ilmvac Products
Click to view available options
Diameter (Metric) Outside:
16 mm
25 mm
40 mm
Conditionnement:
Each
Spécification
Spécification
| Diameter (Metric) Outside | 40 mm |
| For Use With | Separator 40 DN |
| Material | Aluminum |
| Quantity | 1 Each |
| Type | KF Inner Centering |