Learn More
Brady™ Aluminum: ISO Safety Sign - Wear head protection
Sign compliant to ISO 7010.
Brand:  Brady™ 819920
 View more versions of this product
Product Code. 15614527
  • € 36.80 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Description
  • Aluminum
  • For indoor and outdoor use
Specifications
Specifications
ISO 7010
1 Ea.
Indoor and Outdoor
-40°C to 140°C
Global
Aluminum
315 mm
1.016 mm
Round
White on Blue