missing translation for 'onlineSavingsMsg'
Learn More
Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals Product Code.: 11403760

Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals

Product Code. 11403760
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11403760

Brand: Thermo Scientific Alfa Aesar 089350.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Aluminum di(isopropoxide)acetoacetic ester chelate is used as a gel ink. It finds application in paint to give better drying, luster and hardness, and improve the performance as surface wrinkling. It is also used as a crosslinking agent in epoxy, alkyd and polyester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14782-75-3
Molecular Formula C12H23AlO5
MDL Number MFCD00135521

Specifications

Density 1.05 g/mL
Flash Point 50°C (122°F)
Odor Alcohol-like
Quantity 500 g
UN Number UN1993
Sensitivity Moisture sensitive
Formula Weight 274.3
Percent Purity ≥9.6%
Physical Form Liquid
Chemical Name or Material Aluminum di (isopropoxide)acetoacetic ester chelate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ08L012 Certificate TypeCertificate of Analysis Date09/04/2025 Catalog Number089350.22,089350.36
Lot NumberN13L026 Certificate TypeCertificate of Analysis Date17/02/2025 Catalog Number089350.22,089350.36
Lot NumberM20I034 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number089350.22,089350.36
Lot NumberK12M33 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089350.22,089350.36
Lot NumberG10L05 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089350.22,089350.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum di(isopropoxide)acetoacetic ester chelate, Al 9.6% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.