Learn More
3M™ E-A-R™ E-A-Rsoft FX Earplugs
73.10 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Corded:
Corded
Uncorded
Description
- Highest available attenuation rated earplug at 39dB
- Smooth tapered finish makes fitting easier and helps form a comfortable fit
- Flange shape, when rolled down, serves as a very effective insertion device
- Ideal for those looking for higher attenuation
Specifications
Color | Neon Green |
Quantity | 200/Pk. |
Corded | Corded |
Product Type | Ear Plugs |
Style | Roll-down |