Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Click to view available options
Model:
Model A2B2E2K2RD
Model A2B2P3RD
Model ABEK2P3RD
Model AXB2P3
Model HgP3
Unit Size:
Each
Description
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Specifications
| Cartridge Type | Canister |
| Certifications/Compliance | E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7549 A/12/23 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7549 A/12/23 PSA, Certifications- EN 14387, EN 12941, lockedHON filter |
| Connection Type | RD40 |
| Material | Aluminum alloy with PE cover activated and impregnated charcoal |
| Product Type | ALU RD40 A2B2E2K2 |
| For Use With (Application) | Respiratory protection against many hazardous gases, vapors and/or particulates in many applications |
| For Use With (Equipment) | Full-face mask equipped with an Rd40 connector |
| Model | Model A2B2E2K2RD |
| Quantity | 1 Ea. |