Få mere at vide
Få mere at vide
Sperian™ Honeywell™ Aluminum RD40 Filters
Klik for at se tilgængelige muligheder
Model:
Model A2B2E2K2RD
Model A2B2P3RD
Model ABEK2P3RD
Model AXB2P3
Model HgP3
Pakningsstørrelse:
Each
Beskrivelse
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Tekniske data
Tekniske data
| Cartridge Type | Canister |
| Certifications/Compliance | E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7549 A/12/23 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7549 A/12/23 PSA, Certifications- EN 14387, EN 148, EN 12941, HON filte |
| Connection Type | RD40 |
| Material | Aluminum alloy with PE cover activated and impregnated charcoal |
| Product Type | ALU RD40 A2B2E2K2P3 |
| For Use With (Application) | Respiratory protection against ammonia and ammonia derivatives, solid and liquid particles |
| For Use With (Equipment) | Full-face mask equipped with an Rd40 connector |
| Model | Model ABEK2P3RD |
| Quantity | 1 Ea. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion