Learn More
Fisherbrand™ Aluminum Scaffolding Tubes
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Aluminum Scaffolding Tubes are manufactured from a durable aluminum alloy.
Brand: Fisherbrand™ 2121/2
Description
- Approx. 13mm diameter
- Available in two different lengths
- Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Aluminum Alloy | |
2 m | |
Tube |
Product Content Correction
Product Title
Fisherbrand™ Aluminum Scaffolding Tubes > Length: 2m