Learn More
Fisherbrand™ Plane Swivel Scaffolding Connectors
Suitable for making triangular or square frames for supporting large flanged items such as heating mantles and funnels. Fisherbrand™ Plane Swivel Scaffolding Connectors are provided with a steel strap.
Brand: Fisherbrand™
Additional Details : Weight : 0.20000kg
Description
- With thumbscrews for hand tightening
- For connecting two rods or tubes which converge at any angle
Specifications
Aluminum | |
Tripod Connection | |
Pivotal Plan 2-Way |